Få mere at vide
Novacyt - Microgen Bioproducts Latex Agglutination Test: Streptococcus A, B, C, D, F and G
Shop alle Novacyt Group produkterBeskrivelse
- Specific tests available for a variety of infection detections.
- Rapid (2 minute), simple, sensitive, and specific latex agglutination test for the identification and confirmation of infecting colonies.
- Can be used to obtain rapid, presumptive-positive results and provide an early indication of possible production process failure.
- Can be used to screen colonies before full biochemical identification and can replace traditional agglutinating antibody methods.
Tekniske data
Tekniske data
| Indhold og opbevaring | 2°C to 8°C |
| Detekterbare analytter | Streptococci (Group A, B, C, D, F, G) |
| Type | Latex Agglutination Test |
| Mængde | 50 tests |
| Test tid | 2 min. |