Få mere at vide
Larixol, TRC
Artikelnummer. 30483209
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30483209 50 mg
30468702 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30483209 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17595050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1438-66-0
Molekylær formel C20H34O2
Molekylvægt (g/mol) 306.48
Synonym (alphaS,1S,4S,4aS,8aR)-alpha-Ethenyldecahydro-4-hydroxy-alpha,5,5,8a-tetramethyl-2-methylene-1-naphthalenepropanol, [1S-[1alpha(R*),4beta,4abeta,8aalpha]]-alpha-Ethenyldecahydro-4-hydroxy-alpha,5,5,8a-tetramethyl-2-methylene-1-naphthalenepropanol, (+)-Larixol
IUPAC navn (1S,4S,4aR,8aS)-4-[(3S)-3-hydroxy-3-methylpent-4-enyl]-4a,8,8-trimethyl-3-methylidene-2,4,5,6,7,8a-hexahydro-1H-naphthalen-1-ol
SMIL C[C@](O)(CC[C@H]1C(=C)C[C@H](O)[C@H]2C(C)(C)CCC[C@]12C)C=C
Tekniske data
Kemisk navn eller materiale Larixol
Mængde 50 mg
InChI formel InChI=1S/C20H34O2/c1-7-19(5,22)12-9-15-14(2)13-16(21)17-18(3,4)10-8-11-20(15,17)6/h7,15-17,21-22H,1-2,8-13H2,3-6H3/t15-,16-,17-,19+,20+/m0/s1
Formel vægt 306.2559