Få mere at vide
GA International Large Round Cryogenic Writable Labels
Beskrivelse
- Strong adhesive ensures labels won't wrinkle or peel during long-term storage in -80°C freezers, liquid nitrogen and dry ice
- Available in a wide range of colors, for color coding and easy identification
- Small round labels are ideal for Eppendorf tube caps and vials
- Available in a variety of colors
- Writable with permanent cryo markers
Tekniske data
Tekniske data
| Klæbende type | Permanent |
| Farve | Violet |
| Slutte | Matte |
| Format | Roll |
| Materiale | Thermoplastic Film |
| Produkttype | Label |
| Sterilitet | Non-sterile |
| Etiketter pr. række | 2 Labels |
| Mængde | 1100/Pk. |
| Form | Round |
| Vis mere |