Learn More
Hamilton™ Stainless Steel Large Hub Removable Custom Needles
Can be used with removable (RN)-style syringes that are 250ÎĽL and larger.
Brand:  Hamilton™ 7806-10
Product Code. 13204219
  • 1515.00 DKK / Pack of 6
Description
  • Available in various gauges
  • Provided in packs of 6
  • Custom point style and lengths
  • Autoclavable
  • For large hubs
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Hamilton™ Stainless Steel Large Hub Removable Custom Needles > Gauge: 20G; Needle Point Style: 2, 3, or 4