Få mere at vide
MedChemExpress Larazotide acetate
Artikelnummer. 16432946
Encompass_Preferred
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
16432946 25 mg
1 options
Artikelnummer. 16432946 Leverandør MedChemExpress Leverandørnr. HY106268A
MedChemExpress Larazotide acetate is a synthetic peptide. Larazotide acetate acts as a tight junction regulator and reverses leaky junctions to their normally closed state.
Tekniske data
Renhed 98.0%
CAS 881851-50-9
Indhold og opbevaring Sealed storage, away from moisture∣Powder -80°C 2 years, -20°C 1 year∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Til brug med (applikation) COVID-19-immunoregulation
Molekylvægt (g/mol) 785.89 g/mol
Produkttype Peptides
Mængde 25 mg
Forsendelsestilstand Room temperature
Opløselighed H2O : 16.67 mg/mL (21.21 mM∣Need ultrasonic) ∣DMSO : 3.2 mg/mL (4.07 mM∣Need ultrasonic and warming)