Få mere at vide
(+)-Lappaconitine >90%, TRC
Artikelnummer. 30480092
Skift visning
Mængde:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30480092 500 mg
30465092 250 mg
30477303 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30480092 Leverandør Toronto Research Chemicals Leverandørnr. TRCL175850500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 32854-75-4
Molekylær formel C32 H44 N2 O8
Molekylvægt (g/mol) 584.7
Synonym (+)-Lappaconitine, Acetyl-10-deoxysepaconitine
SMIL CCN1C[C@@]2(CC[C@H](OC)C34[C@@H]2CC(C13)[C@@]5(O)C[C@H](OC)[C@H]6C[C@@H]4[C@]5(O)[C@H]6OC)OC(=O)c7ccccc7NC(=O)C
Tekniske data
Kemisk navn eller materiale Lappaconitine
Mængde 500 mg
InChI formel InChI=1S/C32H44N2O8/c1-6-34-16-29(42-28(36)18-9-7-8-10-21(18)33-17(2)35)12-11-25(40-4)31-23(29)14-20(26(31)34)30(37)15-22(39-3)19-13-24(31)32(30,38)27(19)41-5/h7-10,19-20,22-27,37-38H,6,11-16H2,1-5H3,(H,33,35)/t19-,20?,22+,23-,24+,25+,26?,27+,29-,30+,31?,32+/m1/s1
Formel vægt 584.3098