Learn More
LanthaScreen™ Tb-PY20 Antibody Kit
LanthaScreen™ Tb-PY20 Antibody Kit
6140.00 DKK - 95500.00 DKK
Specifications
Molecular Weight of Antigen | 150kDa |
---|---|
Packaging | 1 Kit(s) |
Primary or Secondary | Primary |
Content And Storage | Terbium-labeled anti-pY20 antibody in HEPES buffered saline. Store at -20°C. |
Conjugate | Tb (Terbium)/FITC/Lanthanide Chelates |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11553457
|
Invitrogen
PV3552 |
25 μg |
6140.00 DKK
25µg |
Estimated Shipment: 17-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10042672
|
Invitrogen
PV3553 |
1 mg |
95500.00 DKK
1mg |
Estimated Shipment: 22-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
A general reagent to monitor tyrosine kinase activity, the pY20 antibody binds to phosphorylated tyrosines within a peptide or physiological protein substrate.For use in combination with Fluorescein-poly-GT (PV3610) or Fluorescein-poly-GAT (PV3611) to evaluate inhibitors in medium to high throughput screening applications.
Specifications
150kDa | |
Primary | |
Tb (Terbium)/FITC/Lanthanide Chelates |
1 Kit(s) | |
Terbium-labeled anti-pY20 antibody in HEPES buffered saline. Store at -20°C. | |
RUO, RUO |
For Research Use Only. Not for use in diagnostic procedures.