Learn More
LanthaScreen™ Tb-DUB Substrate
LanthaScreen™ Tb-DUB Substrate
Brand:  Invitrogen PV4443
Additional Details : Weight : 0.00900kg
 View more versions of this product
Product Code. 11505046
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
Deubiquitinating enzymes (DUBs) play housekeeping functions by maintaining pools of active ubiquitin, and regulatory function by rescuing specific target proteins from degradation by the proteasome. The LanthaScreen™ Tb-DUB Substrate is a general substrate for assaying DUB activity in a high throughput assay format.
Specifications
Specifications
10μg
10μg
LanthaScreen™ Tb-DUB Substrate, 10μg, Play Housekeeping Functions by Maintaining Pools of Active Ubiquitin and Regulatory Function by Rescuing Specific Target Proteins from Degradation by the Proteasome, It is a General Substrate For Assaying DUB Activity in a High throughput Assay Format
LanthaScreen™ Tb-DUB Substrate
Pharma and biopharma, target and lead identification and validation, drug discovery and development, proteasome biology, biochemical proteasome assays, epigenetics assays for drug discovery
Deconjugating enzyme substrate
42.8kDa
–80°C