missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lanthanium for AAS 1g/L HNO3 matrix, Fisher Chemical™
Click to view available options
Quantity:
500 mL
Packaging:
Bottle
Specifications
Concentration | 1g/L ±0.002g/L |
CAS | 7697-37-2 |
Packaging | Bottle |
Quantity | 500 mL |
Type | Flame Single-element Standard Solution |
Molecular Formula | HNO3 |
MDL Number | MFCD00011349 |
InChI Key | GRYLNZFGIOXLOG-UHFFFAOYSA-N |
SMILES | O[N+]([O-])=O |
IUPAC Name | nitric acid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'