Få mere at vide
Lansoprazole Sulfone-d4, TRC
Artikelnummer. 30482650
Skift visning
Mængde:
2.5 mg
25 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30482650 2.5 mg 2.5mg
30482178 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30482650 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1750272.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1184999-77-6
Molekylær formel C16H10D4F3N3O3S
Molekylvægt (g/mol) 389.38
Synonym 2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfonyl]-1H-benzimidazole-d4, AG 1813-d4
SMIL CC(C(OCC(F)(F)F)=CC=N1)=C1CS(C(N2)=NC3=C2C([2H])=C([2H])C([2H])=C3[2H])(=O)=O
Specifica
Kemisk navn eller materiale Lansoprazole Sulfone-d4
Mængde 2.5 mg
InChI formel InChI=1S/C16H14F3N3O3S/c1-10-13(20-7-6-14(10)25-9-16(17,18)19)8-26(23,24)15-21-11-4-2-3-5-12(11)22-15/h2-7H,8-9H2,1H3,(H,21,22)/i2D,3D,4D,5D
Titolo del prodotto
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.