Få mere at vide
Lansoprazole Sulfide N-Oxide, TRC
Artikelnummer. 17992645
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17992645 50 mg
17930875 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17992645 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17504550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 163119-30-0
Molekylær formel C16 H14 F3 N3 O2 S
Molekylvægt (g/mol) 369.36
Synonym 2-[[[3-Methyl-1-oxido-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]thio]-1H-benzimidazole, 1H-Benzimidazole, 2-[[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]thio]-, N-oxide, 2-[[[4-(2,2,2-Trifluoroethoxy)-3-methyl-1-oxopyridin-2-yl]methyl]sulfanyl]-1H-benzimidazole, Lansoprazole Sulfide N-Oxide
IUPAC navn 2-[[3-methyl-1-oxido-4-(2,2,2-trifluoroethoxy)pyridin-1-ium-2-yl]methylsulfanyl]-1H-benzimidazole
SMIL Cc1c(OCC(F)(F)F)cc[n+]([O-])c1CSc2nc3ccccc3[nH]2
Tekniske data
Kemisk navn eller materiale Lansoprazole Sulfide N-Oxide
Mængde 50 mg
InChI formel InChI=1S/C16H14F3N3O2S/c1-10-13(22(23)7-6-14(10)24-9-16(17,18)19)8-25-15-20-11-4-2-3-5-12(11)21-15/h2-7H,8-9H2,1H3,(H,20,21)
Formel vægt 369.0759