Få mere at vide
Lansoprazole Sulfide-d4, TRC
Artikelnummer. 30477062
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30477062 25 mg
30469829 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30477062 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17502225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1216682-38-0
Molekylær formel C16H10D4F3N3OS
Molekylvægt (g/mol) 357.39
Synonym 2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]thio]-1H-benzimidazole-d4, AG 1777-d4, H 225/18-d4, K 1252-d4
SMIL [2H]c1c([2H])c([2H])c2[nH]c(SCc3nccc(OCC(F)(F)F)c3C)nc2c1[2H]
Tekniske data
Kemisk navn eller materiale Lansoprazole Sulfide-d4
Mængde 25 mg
InChI formel InChI=1S/C16H14F3N3OS/c1-10-13(20-7-6-14(10)23-9-16(17,18)19)8-24-15-21-11-4-2-3-5-12(11)22-15/h2-7H,8-9H2,1H3,(H,21,22)/i2D,3D,4D,5D