Få mere at vide
Lansoprazole Sulfide-d4, TRC
Artikelnummer. 30469829
Skift visning
Mængde:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30469829 2.5 mg
30477062 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30469829 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1750222.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1216682-38-0
Molekylær formel C16H10D4F3N3OS
Molekylvægt (g/mol) 357.39
Synonym 2-[[[3-Methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]thio]-1H-benzimidazole-d4, AG 1777-d4, H 225/18-d4, K 1252-d4
SMIL [2H]c1c([2H])c([2H])c2[nH]c(SCc3nccc(OCC(F)(F)F)c3C)nc2c1[2H]
Tekniske data
Kemisk navn eller materiale Lansoprazole Sulfide-d4
Mængde 2.5 mg
InChI formel InChI=1S/C16H14F3N3OS/c1-10-13(20-7-6-14(10)23-9-16(17,18)19)8-24-15-21-11-4-2-3-5-12(11)22-15/h2-7H,8-9H2,1H3,(H,21,22)/i2D,3D,4D,5D