Få mere at vide
Lansoprazole-d4, TRC
Artikelnummer. 30464790
Skift visning
Mængde:
2.5 mg
5 mg
25 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30464790 2.5 mg 2.5mg
30465856 25 mg -
30481719 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464790 Leverandør Toronto Research Chemicals Leverandørnr. TRCL1750022.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 934294-22-1
Molekylær formel C16 2H4 H10 F3 N3 O2 S
Molekylvægt (g/mol) 373.39
Synonym 1H-Benzimidazole-4,5,6,7-d4, 2-[[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfinyl]-, Lansoprazole-D4, Lansoprazole-D4 (benzimidazole-4,5,6,7-D4)
IUPAC navn 4,5,6,7-tetradeuterio-2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1H-benzimidazole
SMIL [2H]c1c([2H])c([2H])c2[nH]c(nc2c1[2H])S(=O)Cc3nccc(OCC(F)(F)F)c3C
Tekniske data
Kemisk navn eller materiale Lansoprazole-d4
Renhedsgrad noter HPLC
Mængde 2.5 mg
InChI formel InChI=1S/C16H14F3N3O2S/c1-10-13(20-7-6-14(10)24-9-16(17,18)19)8-25(23)15-21-11-4-2-3-5-12(11)22-15/h2-7H,8-9H2,1H3,(H,21,22)/i2D,3D,4D,5D
Formel vægt 373.101
Procent renhed >95%