Få mere at vide
Lansoprazole-d4 Sulfone N-Oxide, TRC
Artikelnummer. 30466906
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30466906 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30466906 Leverandør Toronto Research Chemicals Leverandørnr. TRCL17503210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lansoprazole-d4 Sulfone N-Oxide
CAS 1190009-70-1
Molekylær formel C16H10D4F3N3O4S
Mængde 10 mg
Synonym 2-[[[3-Methyl-1-oxido-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfonyl]-1H-benzimidazole-d4
InChI formel InChI=1S/C16H14F3N3O4S/c1-10-13(22(23)7-6-14(10)26-9-16(17,18)19)8-27(24,25)15-20-11-4-2-3-5-12(11)21-15/h2-7H,8-9H2,1H3,(H,20,21)/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c2[nH]c(nc2c1[2H])S(=O)(=O)Cc3c(C)c(OCC(F)(F)F)ccn3=O
Molekylvægt (g/mol) 405.38