Få mere at vide
Lansoprazole-d4 (benzimidazole-4,5,6,7-d4), CDN
Artikelnummer. 30328437
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328437 0.005 g
30319657 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30328437 Leverandør CDN Leverandørnr. CDND60960.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lansoprazole-d4
CAS 934294-22-1
Molekylær formel C16 2H4 H10 F3 N3 O2 S
Mængde 0.005 g
Synonym 1 H-Benzimidazole-4,5,6,7-d4, 2-[[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfinyl]-, Lansoprazole-D4, Lansoprazole-D4 (benzimidazole-4,5,6,7-D4)
InChI formel InChI=1 S/C16H14F3N3O2S/c1-10-13(20-7-6-14(10)24-9-16(17,18)19)8-25(23)15-21-11-4-2-3-5-12(11)22-15/h2-7 H,8-9H2,1H3,(H,21,22)/i2D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c2[nH]c(nc2c1[2 H])S(=O)Cc3nccc(OCC(F)(F)F)c3C
IUPAC navn 4,5,6,7-tetradeuterio-2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1 H-benzimidazole
Molekylvægt (g/mol) 373.39
Formel vægt 373.101 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity