Learn More
Logistik™ Lancer-Style Chemical Splash Laboratory Coats
Product Code. 12834382
Change view
Click to view available options
Size:
Small
Large
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
12834382 Small Each
12814382 Large Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12834382 Supplier Logistik™ Supplier No. 849AE2WH92CM
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Denna artikel kan inte returneras. Se returpolicy
Provide the answer to full frontal protection with Logistik™ Lancer-Style Chemical Splash Laboratory Coats The wrap-over style Lancer front coat helps repel liquid chemicals and provides the wearer with an excellent level of protection since the use of chemicals in a laboratory can prove a constant hazard to standard style open neck coats.
  • Full frontal protection
  • Chem-Tech material
  • Fluorochemical liquid repellent
Specifikationer
Product Type General Purpose Lab Coat
Size Small
Gender Unisex
Material Polyester, Carbon
Color White
Cuff Style Stud-fastened
Disposable Reusable
Sterility Non-sterile
Style Lancer
No. of Pockets 2
Collar Type Open
Finish Fluorochemical Liquid Repellent
Size (Metric) Chest 91.44 to 96.52 cm
Certifications/Compliance CE EN 13034
Length (Metric) 92 cm
Construction 65% Polyester, 35% Cotton
Visa mer Visa mindre
Produkttitel
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.