Få mere at vide
Cole Parmer LaMotte Sanitizer Strength QAC Test Strips
Shop alle Cole-Parmer produkter
Mængde:
100 strimler/sæt
Pakningsstørrelse:
1 sæt
Description
LaMotte Sanitizer Strength QAC Test Strips are a quick test for sanitizer strength.
- Compare color change to included chart for easy-to-read results.
- Strips and color chart come supplied in a waterproof plastic vial.
- Include a dessiccant cap to ensure longer life.
- Measurement Points: 50, 100, 200 and 400 ppm
Specifications
Specifications
| Format | Strip |
| Mængde | 100 strimler/sæt |
| Type | Chlorine and pH Test Kits and Test Strips |