Learn More
Brady™ Laminated Polyester Unauthorized Access Prohibited Signs
Brand: Brady™ 250019
19.11 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
This item is not returnable.
View return policy
Specifications
PIC 209-DIA 100-B7541 (BRANDED) | |
100 mm | |
Round |
Promotions
-
Multiple Manufacturers
Everyday Lab Essentials
From wipes to waste bins, labels to liners – save on them all.Promo Code: 24132Expires: 2025-08-24
New Offer Shop Now
Product Content Correction
Product Title
Brady™ Laminated Polyester Unauthorized Access Prohibited Signs > Dia.: 100mm
Spot an opportunity for improvement?Share a Content Correction