Learn More
Brady™ Laminated Polyester ADR 3b Flammable Liquid Transport Signs
Product Code. 11780244 Shop All Brady Products
Change view
Click to view available options
Height (Metric):
100 mm
200 mm
297 mm
Width (Metric):
100.00 mm
200.00 mm
297.00 mm
Quantity:
1 Ea.
250/Pk.
Unit Size:
1 roll
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Width (Metric) Quantity Unit Size
11780244 100 mm 100.00 mm 250/Pk. 1 roll
11740214 200 mm 200.00 mm 1 Ea. Each
11730214 100 mm 100.00 mm 1 Ea. Each
11750214 297 mm 297.00 mm 1 Ea. Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 11780244 Supplier Brady™ Supplier No. 223604
  • ONLINE EXCLUSIVE PRICE
    3055.00 DKK
    2846.57 DKK / 1 ruller
    Spare 208.43 DKK
    6% Slukket
Forventet forsendelse: 10-12-2026
Specifications
Alternate Part Number ADR 3BRL-100*100-B7541
Finish Gloss
Material Laminated Polyester
Quantity 250/Pk.
Adhesive Type Permanent Acrylic
Height (Metric) 100 mm
Thickness 0.099 mm
Width (Metric) 100.00 mm
Temperature Range (Metric) -40 °C-110 °C
Shape Diamond
Displayed Text 3
Color White on Red