Få mere at vide
Lake Red CBA (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Lake Red CBA |
| CAS | 5160-02-1 |
| Molekylær formel | 2[C17 H12 Cl N2 O4 S] . Ba |
| Mængde | 250 mg |
| Synonym | Pigment Red 53:1, Benzenesulfonic acid, 5-chloro-2-[(2-hydroxy-1-naphthalenyl)azo]-4-methyl-, barium salt (2:1) (9CI), C.I. Pigment Red 53, Ba salt (7CI), C.I. Pigment Red 53, barium salt (2:1) (8CI), 1860 Red, Astro Orange, Atomic Red, Bronze Scarlet CA, Bronze Scarlet CBA, Bronze Scarlet CTA, C.I. 15585:1, C.I. PR 53:1, C.I. Pigment Red 53:1, Carnation Red, D and C Red No. 9, D&C Red No. 9, Desert Red, Eljon Lake Red C, Irgalite Red CBNL, Irgalite Red LCB, Japan Red 204, Japan Red No. 204, Jinguanghong, Jinguanghong C, Lake Red C, Lake Red C 18287, Lake Red C 21245, Lake Red C 27200, Lake Red C 27217, Lake Red C 27218, Lake Red C-RLC 232, Lake Red CBA, Lake Red GB, Lake Red Toner LCLL, Lake Red ZhB, Latexol Scarlet R, Lionol Red C-FG 3100, PR 53:1, Permanent Lake Red LC, Permanent Lake Red LCLL, Pigment Lake Red BFC, Pigment Lake Red LC, Pigment Red 53:1, Red 6011, Red Lake C, Red No. 204, Red ZhB, Scarlet LC, Segnale Red LCG, Segnale Red LCL, Sumikaprint Red C-O, Symuler Lake Red C |
| InChI formel | InChI=1S/2C17H13ClN2O4S.Ba/c2*1-10-8-14(16(9-13(10)18)25(22,23)24)19-20-17-12-5-3-2-4-11(12)6-7-15(17)21;/h2*2-9,21H,1H3,(H,22,23,24);/q;;+2/p-2 |
| SMIL | [Ba+2].Cc1cc(N=Nc2c(O)ccc3ccccc23)c(cc1Cl)S(=O)(=O)[O-].Cc4cc(N=Nc5c(O)ccc6ccccc56)c(cc4Cl)S(=O)(=O)[O-] |
| IUPAC navn | barium(2+);5-chloro-2-[(E)-(2-hydroxynaphthalen-1-yl)diazenyl]-4-methylbenzenesulfonate |
| Molekylvægt (g/mol) | 888.93 |
| Formel vægt | 887.9465 |