missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LAF4 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06186FR
Additional Details : Weight : 0.00970kg
Description
LAF4 Polyclonal specifically detects LAF4 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
LAF4 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
AF4/FMR2 family, member 3 | |
Produced in rabbits immunized with E. coli-derived Human LAF4 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
DyLight 680 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
3899 | |
Store at 4°C in the dark. | |
IgG |