missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
LADMAC (mouse bone marrow, producing colony stimulating factor-1) Whole cell lysate, Non-denatured; Abnova
Beskrivelse
- Tissue: Bone marrow
- Host: Mouse
- Lysis buffer: Modified RIPA lysis buffer: 50mM Tris-HCl pH 7.4, 150mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF
- Storage buffer: In modified RIPA lysis buffer
Western Blotting, Immunoprecipitation
Tekniske data
Tekniske data
| Til brug med (applikation) | Immunoprecipitation, Western Blot |
| Væv | Bone Marrow |
| Oversigt | Whole cell lysate (non-denatured) |
| Lyseringsbuffer | Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. |
| Fremstillingsmetode | Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). |
| Kvalitetskontrol test | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Mængde | 200 μg |
| Opbevaringsbuffer | In modified RIPA Lysis Buffer. |
| Værtsarter | Mouse |
| Indhold og opbevaring | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?