missing translation for 'onlineSavingsMsg'
Learn More

Lactulose, MP Biomedicals™

Catalog Number p-7083062
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
4618-18-2
C12H22O11
342.297
JCQLYHFGKNRPGE-DJYZFUSFSA-N
lactulose
131839611
(2S,3S,4S,5R,6R)-2-[(2R,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2C(OC(C2O)(CO)O)CO)O)O)O)O
This item is not returnable. View return policy
4618-18-2
C12H22O11
342.297
JCQLYHFGKNRPGE-DJYZFUSFSA-N
lactulose
131839611
(2S,3S,4S,5R,6R)-2-[(2R,4S,5R)-4,5-dihydroxy-2,5-bis(hydroxymethyl)oxolan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C(C1C(C(C(C(O1)OC2C(OC(C2O)(CO)O)CO)O)O)O)O

Used in the treatment of chronic constipation, SIBO and hepatic encephalopathy. MP Biomedicals' Lactulose is a synthetic, non-digestible disaccharide (double-sugar) formed from one molecule each of fructose and galactose (monosaccharides). It is also used

pH 5.5 to 6.5 (1% aq. soln.)
Quantity 100 g
Solubility Information Soluble in 5% of water (colorless)
Optical Rotation −45 to −52° (c=5, water, 24 hrs)
Formula Weight 342.3
Percent Purity ∼98%
Physical Form Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'