Få mere at vide
Lactose Olamine Adduct (>80%), TRC
Artikelnummer. 30471675
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30471675 50 mg
30465602 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30471675 Leverandør Toronto Research Chemicals Leverandørnr. TRCL11402050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 193979-22-5
Molekylær formel C14 H27 N O11
Molekylvægt (g/mol) 385.364
Synonym Eltrombopag Olamine Impurity
IUPAC navn (2S,3R,4S,5R,6R)-2-[(2R,3S,4R,5R,6S)-4,5-dihydroxy-6-(2-hydroxyethylamino)-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMIL OCCN[C@H]1O[C@H](CO)[C@@H](O[C@@H]2O[C@H](CO)[C@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O
Tekniske data
Kemisk navn eller materiale Lactose Olamine Adduct
Mængde 50 mg
InChI formel InChI=1S/C14H27NO11/c16-2-1-15-13-10(22)9(21)12(6(4-18)24-13)26-14-11(23)8(20)7(19)5(3-17)25-14/h5-23H,1-4H2/t5-,6-,7+,8+,9-,10-,11-,12-,13+,14+/m1/s1
Formel vægt 385.158