missing translation for 'onlineSavingsMsg'
Få mere at vide

Lactal Hexaacetate, TRC

Artikelnummer. 30475536
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30475536 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30475536 Leverandør Toronto Research Chemicals Leverandørnr. TRCL112500500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Lactal Hexaacetate
CAS 51450-24-9
Molekylær formel C24 H32 O15
Mængde 500 mg
InChI formel InChI=1S/C24H32O15/c1-11(25)32-9-18-20(17(7-8-31-18)34-13(3)27)39-24-23(37-16(6)30)22(36-15(5)29)21(35-14(4)28)19(38-24)10-33-12(2)26/h7-8,17-24H,9-10H2,1-6H3/t17-,18-,19-,20+,21+,22+,23-,24+/m1/s1
SMIL CC(=O)OC[C@H]1OC=C[C@@H](OC(=O)C)[C@@H]1O[C@@H]2O[C@H](COC(=O)C)[C@H](OC(=O)C)[C@H](OC(=O)C)[C@H]2OC(=O)C
IUPAC navn [(2R,3S,4R)-4-acetyloxy-3-[(2S,3R,4S,5S,6R)-3,4,5-triacetyloxy-6-(acetyloxymethyl)oxan-2-yl]oxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
Molekylvægt (g/mol) 560.5
Formel vægt 560.1741
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.