missing translation for 'onlineSavingsMsg'
Learn More

Lacosamide, TRC

Product Code. 30470238
Change view
Click to view available options
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30470238 100 mg
30479452 10 mg
30474357 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30470238 Supplier Toronto Research Chemicals Supplier No. TRCL098500100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175481-36-4
Molekylær formel C13 H18 N2 O3
Molekylvægt (g/mol) 250.29
Synonym (2R)-2-(Acetylamino)-3-methoxy-N-(phenylmethyl)-propanamide, Propanamide, 2-(acetylamino)-3-methoxy-N-(phenylmethyl)-, (R)-, (2R)-2-(Acetylamino)-3-methoxy-N-(phenylmethyl)propanamide, (2R)-2-Acetamido-N-benzyl-3-methoxypropanamide, (R)-2-Acetamido-N-benzyl-3-methoxypropionamide, ADD 243037, Erlosamide, Harkoseride, Lacosamide, Lacoset, Lacoset 50, SPM 927, Vimpat
IUPAC navn (2R)-2-acetamido-N-benzyl-3-methoxypropanamide
SMIL COC[C@@H](NC(=O)C)C(=O)NCc1ccccc1

Specifications

Kemisk navn eller materiale Lacosamide
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C13H18N2O3/c1-10(16)15-12(9-18-2)13(17)14-8-11-6-4-3-5-7-11/h3-7,12H,8-9H2,1-2H3,(H,14,17)(H,15,16)/t12-/m1/s1
Formel vægt 250.1317
Procent renhed >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.