Få mere at vide
Lacidipine-d10, TRC
Artikelnummer. 30464794
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30464794 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30464794 Leverandør Toronto Research Chemicals Leverandørnr. TRCL09800210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Lacidipine-d10
CAS 1185245-62-8
Molekylær formel C26H23D10NO6
Mængde 10 mg
Synonym (E)-4-[2-[3-(1,1-Dimethylethoxy)-3-oxo-1-propenyl]phenyl]-1,4-dihydro-2,6-dimethyl-3,5-pyridinedicarboxylic Acid Diethyl Ester-d10, GR-43659X-d10, GX-1048-d10, Caldine-d10, Lacipil-d10, Lacirex-d10, Motens-d10
InChI formel InChI=1S/C26H33NO6/c1-8-31-24(29)21-16(3)27-17(4)22(25(30)32-9-2)23(21)19-13-11-10-12-18(19)14-15-20(28)33-26(5,6)7/h10-15,23,27H,8-9H2,1-7H3/b15-14+/i1D3,2D3,8D2,9D2
SMIL CC1=C(C(OC([2H])([2H])C([2H])([2H])[2H])=O)C(C2=CC=CC=C2/C=C/C(OC(C)(C)C)=O)C(C(OC([2H])([2H])C([2H])([2H])[2H])=O)=C(C)N1
Molekylvægt (g/mol) 465.6