Få mere at vide
Laccaic Acid (Technical Grade), TRC
Artikelnummer. 30483972
Skift visning
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30483972 2.5 g 2.5g
30475072 500 mg -
30466357 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30483972 Leverandør Toronto Research Chemicals Leverandørnr. TRCL3001302.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 60687-93-6
Molekylær formel C91H62N2O44
Molekylvægt (g/mol) 1887.45
Synonym C.I. Natural Red 25, C.I. 75450, Lac, Lac dye, Lac lake, Natural Red 25, RK Color 7LA200L, Red Lac, Rhine M, San Red LN, Shiko, Stilacred L 45, Stilacred L 5
SMIL CC1=C2C(=CC(=C1C(=O)O)O)C(=O)C3=CC(=CC(=C3C2=O)O)O.CC(=O)NCCC1=CC(=C(C=C1)O)C2=C(C3=C(C(=C2O)O)C(=O)C4=CC(=C(C(=C4C3=O)C(=O)O)C(=O)O)O)O.C1=CC(=C(C=C1CCO)C2=C(C3=C(C(=C2O)O)C(=O)C4=CC(=C(C(=C4C3=O)C(=O)O)C(=O)O)O)O)O.C1=CC(=C(C=C1CC(C(=O)O)N)C2=C(C3=C(C(=C2O)O)C(=O)C4=CC(=C(C(=C4C3=O)C(=O)O)C(=O)O)O)O)O
Tekniske data
Kemisk navn eller materiale Laccaic Acid (Technical Grade)
Mængde 2.5 g
InChI formel InChI=1S/C26H19NO12.C25H17NO13.C24H16O12.C16H10O7/c1-8(28)27-5-4-9-2-3-12(29)10(6-9)15-22(33)19-18(24(35)23(15)34)20(31)11-7-13(30)16(25(36)37)17(26(38)39)14(11)21(19)32;26-9(23(34)35)4-6-1-2-10(27)7(3-6)13-20(31)17-16(22(33)21(13)32)18(29)8-5-11(28)14(24(36)37)15(25(38)39)12(8)19(17)30;25-4-3-7-1-2-10(26)8(5-7)13-20(30)17-16(22(32)21(13)31)18(28)9-6-11(27)14(23(33)34)15(24(35)36)12(9)19(17)29;1-5-11-8(4-10(19)12(5)16(22)23)14(20)7-2-6(17)3-9(18)13(7)15(11)21/h2-3,6-7,29-30,33-35H,4-5H2,1H3,(H,27,28)(H,36,37)(H,38,39);1-3,5,9,27-28,31-33H,4,26H2,(H,34,35)(H,36,37)(H,38,39);1-2,5-6,25-27,30-32H,3-4H2,(H,33,34)(H,35,36);2-4,17-19H,1H3,(H,22,23)
Formel vægt 1886.27