Få mere at vide
Labplas Sani-Sponge Kit
The simpler solution to surface sampling
Brand: Labplas KSS-61100
Beskrivelse
Designed to collect samples to detect the presence of microbiological contaminations such as Listeria, Salmonella, E. coli, and other food-borne pathogens on almost any surface. Kit is convenient and easy to use, and helps eliminate several time-consuming steps.
- All Labplas SANI-SPONGE TWIRL'EM™ bags come with safety tabs
- Puncture-proof tabs eliminate the sharp points that can cause bag puncture and/or abrasions
- Kits are sterilized using gamma irradiation
- Produced in accordance with HACCP, USDA, ACIA and CFIA requirements
- Available in Dry Sponge or Wet Sponge forms
Dry Sponge
- TWIRL'EM™ Sterile Sampling bag with write-on strip containing a dry biocide-free cellulose sponge (1.5 in. × 3 in. × 0.625 in. when hydrated)
- TWIRL'EM Sterile Sampling bag accompanied by a foil-sealed, pre-moistened cellulose sponge, holding 10mL of neutralizing buffer-sterile
Tekniske data
| 18 oz. | |
| 9 x 4.5 in. (229 x 114mm) | |
| 229 mm | |
| 114 mm |
| Sponge | |
| 9 in. | |
| 4.5 in. |