Få mere at vide
DuraLab Laboratory Trolley Small
Artikelnummer. p-200136276
Skift visning
Antal hylder:
2
3
4
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Antal hylder
30280267 2
30280282 3
30280297 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30280267 Leverandør DuraLab Leverandørnr. DRL0001
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Lightweight and versatile with 250 kg capacity, designed with precision castors and brakes for smooth, safe, and reliable mobility.
  • Constructed from recycled Polypropylene to minimise environmental impact
  • Reinforced with solid steel blocks for added strength and durability
  • Fitted with 4 rubber castors, including 2 with locks, for secure and smooth mobility
  • Crafted from high-grade nylon fibre blended polymers to endure demanding conditions
  • Designed with a 250 kg load capacity for dependable use in laboratories
Tekniske data
Produkttype Double Trolley
Farve Black
Dimensioner (L x B x H) 850 x 480 x 950 mm
Bæreevne (engelsk) 551.15 lb.
Bæreevne (metrisk) 250 kg
Materiale Aluminum, Polymer, High Modulus Rubber, Polypropylene
Antal hylder 2