missing translation for 'onlineSavingsMsg'
Få mere at vide

DuraLab Laboratory Trolley Large

Artikelnummer. p-200136274
Skift visning
Klik for at se tilgængelige muligheder
Antal hylder:
2
3
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Antal hylder
30280262 2
30280264 3
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30280262 Leverandør DuraLab Leverandørnr. DRL0004
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy

Laboratory trolleys are lightweight, versatile, and durable, ensuring smooth mobility and reliable performance in laboratories.

  • Versatile design suitable for various laboratory applications.
  • Lightweight construction with strong load-bearing capacity.
  • Equipped with precision bearing castors, including 2 with brakes, for smooth and safe mobility.
  • Crafted from top-grade nylon fibre blended polymers to withstand rigorous working conditions.
  • Constructed from recycled Polypropylene (PP) to support sustainability and minimise environmental impact.
  • Reinforced with solid steel blocks for enhanced durability.
  • Includes 4 rubber castors (2 with locks) for stability and easy movement.
  • Designed to support a load capacity of up to 250 kg.

Tekniske data

Produkttype Double Trolley
Farve Black
Bæreevne (engelsk) 551.15 lb.
Bæreevne (metrisk) 250 kg
Materiale Aluminum, Polypropylene, Polymer, High Modulus Rubber
Antal hylder 2
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.