Beskrivelse
- Variable-speed heater pump
- EcoDry via patented AutoOpen function
- Optimum cleaning performance
- Optimum wash pressure
- Low consumption values
- Short batch times
- Crevice-free wash cabinet
- Spray arm pressure monitoring
- Spray arm monitoring
- Hot water connection
- De-min. water connection
- Liquid dispensing
Tekniske data
Tekniske data
| Oversigt | Freestanding or built-under |
| Materiale | Stainless Steel |
| Flowhastighed | 500 L/min. |
| Certificeringer/compliance | IP safety class according to EN 60529: IP 21, EN 61010-1, EN 61010-2-040, c CSA us |
| Inkluderer | Hot water connection, de-min. water connection, liquid dispensing, automatic door lock, acoustic end of program signal, integrated water softener |
| Indløbsvandtemperatur (engelsk) Maks. | 93°C |
| Mål (D x B x H) Indvendig | 474/520 x 530 x 520mm |
| Elektriske krav | 2 AC 208V 60HZ |