Få mere at vide
Heinz Herenz Laboratory Dropper Bottles with Narrow Neck
Beskrivelse
- Laboratory dropper bottles
- 500 mL
- CircularLine
- Narrow neck, with long tip and attached red protection cap
- Made from sustainable LDPE with more than 30% made from recycled renewable resources.
Tekniske data
Tekniske data
| Kapacitet (metrisk) | 500 mL |
| Farve | Transparent, Natural |
| Materiale | LDPE |
| Sterilitet | Non-sterile |
| Trådstørrelse (metrisk) | GL32 |
| Lukning inkluderet | Yes |
| Engangs | Reusable |
| Uddannet | No |
| Mund | Narrow, Threaded |
| Form | Round |
| Vis mere |