Få mere at vide
Heinz Herenz Laboratory Dish Set 6 Pieces
Beskrivelse
- Laboratory dish set
- 6 piece.set: 0.5 L - 180 x 230 x 42 mm, 1.5 L - 250 x 310 x 65 mm, 3.0 L - 310 x 370 x 75 mm, 10.0 L - 420 x 520 x 120 mm, 21.0 L - 534 x 634 x 140 mm, 39.0 L - 648 x 846 x 160 mm
- Plastic
Tekniske data
Tekniske data
| Autoklaverbar | Autoclavable |
| Bund form | Flat |
| Farve | White |
| Engangs | Reusable |
| Form | Rectangular |
| Sterilitet | Non-sterile |
| Overfladebehandling | Non-treated |
| Type | Laboratory Dish |
| Højde (metrisk) | 140 mm |