Learn More
SHP Steriltechnik AG Laboklav 135-195 Bucket
Product Code. 31908738
Change view
Click to view available options
For Use With (Equipment):
Laboklav 135, Laboklav 160, Laboklav 195
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
31908738 Laboklav 135, Laboklav 160, Laboklav 195 Each
1 options
Product Code. 31908738 Supplier SHP Steriltechnik AG Supplier No. 30.1500.000.300
Forventet forsendelse: 15-01-2027
Stainless steel bucket for Laboklav 135-195, vertical unit.
  • For vertical units
  • Stainless Steel
  • ø x H (mm): 500 x 300
Specifications
Capacity (Metric) 58 L
For Use With (Equipment) Laboklav 135, Laboklav 160, Laboklav 195
Model Bucket for Laboklav 135-195
Product Type Bucket
Sterility Non-sterile