Få mere at vide
Labetalol EP Impurity B (as Hydrochloride), Mikromol™
Beskrivelse
Labetalol EP Impurity B (as Hydrochloride)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Methyl 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoate Hydrochloride |
| CAS | 33778-93-7 |
| Analyt- eller komponentnavne | Methyl 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoate Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H25 N O4 . Cl H |
| Certificeringer/compliance | ISO 17025 |
| Synonym | Labetalol Hydrochloride Imp. B (EP), Labetalol Imp. B (EP), Methyl 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoate Hydrochloride, Labetalol Hydrochloride Imp. B (EP) as Hydrochloride, Labetalol Hydrochloride Impurity B as Hydrochloride, Labetalol Impurity B as Hydrochloride |
| InChI formel | InChI=1S/C20H25NO4.ClH/c1-14(8-9-15-6-4-3-5-7-15)21-13-19(23)16-10-11-18(22)17(12-16)20(24)25-2;/h3-7,10-12,14,19,21-23H,8-9,13H2,1-2H3;1H |
| SMIL | Cl.COC(=O)c1cc(ccc1O)C(O)CNC(C)CCc2ccccc2 |
| IUPAC navn | methyl 2-hydroxy-5-[1-hydroxy-2-(4-phenylbutan-2-ylamino)ethyl]benzoate;hydrochloride |
| Vis mere |