Få mere at vide
Labetalol 1-carboxylic Acid Methyl Ester, TRC
Artikelnummer. 30465858
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30465858 25 mg
1 options
Artikelnummer. 30465858 Leverandør Toronto Research Chemicals Leverandørnr. TRCL09651525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Labetalol 1-carboxylic Acid Methyl Ester
CAS 802620-01-5
Molekylær formel C20H25NO4
Mængde 25 mg
Synonym Methyl 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoate, 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoic Acid Methyl Ester, 5-[1-Hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]salicylic Acid Methyl Ester, Labetalol Impurity B
InChI formel InChI=1S/C20H25NO4/c1-14(8-9-15-6-4-3-5-7-15)21-13-19(23)16-10-11-18(22)17(12-16)20(24)25-2/h3-7,10-12,14,19,21-23H,8-9,13H2,1-2H3
SMIL CC(CCC1=CC=CC=C1)NCC(C2=CC(=C(C=C2)O)C(=O)OC)O
Molekylvægt (g/mol) 343.42