Learn More
Labconco™ Replacement Prefilter for Protector Downdraft Powder Station
Replacement Prefilter for use in Protector Downdraft Powder Stations.
215.00 DKK - 1845.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
15470412
|
Labconco™
3768901 |
4 ft. Purifier™ horizontal clean benches and 6 ft. Purifier™ horizontal clean benches |
215.00 DKK
Each |
Estimated Shipment: 05-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11781469
|
Labconco™
9067800 |
WaterPro RO Stations |
1845.00 DKK
Each |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Replacement Prefilter for use in Protector Downdraft Powder Stations.
Replacement Prefilter for use with Labconco Protector Downdraft Powder Station forensic enclosures. Made of pleated paper. MERV 8 rating.Specifications
Prefilter | |
Prefilter |