Learn More
Labconco™ Replacement Filters for Benchtop Fume Adsorbers
Replacement filters for use with Labconco Fume Adsorbers and Fume Adsorber-2s.
Brand: Labconco™ 6961301
Additional Details : Weight : 4.00000kg
Description
Replacement Formaldehyde Carbon Filter for use with formaldehyde and gluteraldehyde. 7.5 pounds of impregnated carbon. Adsorption weight 10% (0.75 lb.). Adsorption volume 850 ml of 37% formalin. For use with Labconco Fume Adsorber and Fume Adsorber-2. Fume Adsorber Models 3955201 and 3955221 include one filter; Fume Adsorber-2 Models 3955401 and 3955421 include two filters.Specifications
Carbon | |
11 lb. | |
Filter |
Benchtop Fume Adsorber | |
5 Kg | |
Formaldehyde-formalin |