Learn More
Labconco™ Filters
Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement
5610.00 DKK
Specifications
Efficiency | 99.99% |
---|---|
Filter Type | HEPA |
Product Type | Filter |
For Use With (Application) | Particulate |
Product Code | Brand | Length (English) | Weight (English) | Length (Metric) | Weight (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (English) | Weight (English) | Length (Metric) | Weight (Metric) | Price | Quantity & Availability | ||
10109152
|
Labconco™
3707900 |
10 to 20 in. | 40 lb. | 25.4 to 50.8 cm | 18.1 Kg |
5610.00 DKK
Each |
Estimated Shipment: 09-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |
Description
Carbon-based and HEPA filters for use in Labconco enclosures which require periodic replacement
Replacement HEPA Filter, 99.99% efficient on particles 0.3 micron in size. Not suitable for use with biohazardous materials or volatile organic material. For use with Labconco FilterMate Portable Exhausters, Purifier Class I Enclosures, Purifier Filtered PCR Enclosures, Purifier HEPA Filtered Enclosures, Purifier Vertical Clean Benches, XPert Filtered Balance Stations, XPert and RXPert Filtered Balance Systems.Specifications
99.99% | |
HEPA | |
Filter | |
Particulate |