Learn More
Labconco™ Lids for CentriVap™Cold Traps
Provides corrosion resistance.
Brand: Labconco™ 7386700
Additional Details : Weight : 2.70000kg
Description
Stainless Steel Lid may be installed in place of the acrylic lid on -50 CentriVap Cold Trap when applications involve solvents which attack acrylic. Included on CentriVap Solvent and Proteomic Systems and -84 and -105 CentriVap Cold Traps.Specifications
Stainless Steel | |
Concentrator Cold Trap Lid | |
CentriVap™ concentrator cold trap |