Learn More
Labconco™ HEPA Filter Vacuum Trap w/Tubing
Filter particulates before entry into vacuum pump by adding a Labconco Glove Box In-Line HEPA Filter Relief Bubbler Kit to your system.
Brand: Labconco™ 5441400
Additional Details : Weight : 4.50000kg
Description
- Filters particulates before entry into vacuum pump
- Use w/Atmospure Re-Gen Gas Purifier
Specifications
HEPA | |
12 in. | |
15 in. | |
38.1 cm | |
38.1 cm |
HEPA filter for AtmosPure™ Re-Gen gas purifier | |
15 in. | |
12 x 15 in. (30.48 x 38.1 cm) | |
30.48 cm |