Learn More
Labconco™ HEPA Filter Vacuum Trap w/Tubing
Filter particulates before entry into vacuum pump by adding a Labconco Glove Box In-Line HEPA Filter Relief Bubbler Kit to your system.
Brand:  Labconco™ 5441400
Additional Details : Weight : 4.50000kg
Product Code. 15659256
  • 5960.00 DKK / Each
Estimated Shipment: 08-07-2024
to see stock.
Description
Description
  • Filters particulates before entry into vacuum pump
  • Use w/Atmospure Re-Gen Gas Purifier
Specifications
Specifications
HEPA
12 in.
15 in.
38.1 cm
38.1 cm
HEPA filter for AtmosPure™ Re-Gen gas purifier
15 in.
12 x 15 in. (30.48 x 38.1 cm)
30.48 cm