Få mere at vide
Labconco™ Caster Kits
Replaces fixed feet included with electric and manual hydraulic lift base stands to add mobility
Brand: Labconco™ 3784000
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
- Includes four 4.5 in. dia. (12.7cm) nonmarking polyurethane casters with toe locks and bearings for quiet movement
- Load rating: 250 lb. (113.4kg) per caster
- Installation required