Få mere at vide
Labcon™ Labcon ProtectR™Tubes for Storage and Shipping on Dry Ice
GREENER_CHOICE
Artikelnummer. 30280315 Shop alle Labcon produkter
Skift visning
Sterilitet:
Ikke-steril
Steril
Kapacitet (metrisk):
5 ml
10 ml
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Sterilitet Kapacitet (metrisk)
30280315 Steril 10 ml
30280333 Steril 5 ml
30280318 Ikke-steril 5 ml
30280326 Ikke-steril 10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 30280315 Leverandør Labcon™ Leverandørnr. 38251650009
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Conform to all biological sample shipping requirements. Designed to be CO2 resistant and stop costly damage to molecular and cellular solutions when using dry ice. Airtight and leakproof secure seal ensures consistent sample integrity.
  • CO2 resistant elastomeric sealed caps
  • Certified to prevent acidification from dry ice shipment or storage
  • Certified to IATA/U.S. DOT/UN3373 standards
  • ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
  • Polypropylene USP Class VI resin
  • Centrifuging to 12,500 x G
  • Freezable to -80°C
  • Made in USA
Tekniske data
Certificeringer/compliance ISO 9001:2015, FDA CFR 21 GMP, IATA 95 kPa, UN3373 standard
Lukningsmateriale Polypropylene, Class VI
Kapacitet (metrisk) 10 ml
Engangs Single-use
Materiale Polypropylene
Til brug med Shipping on dry ice
Emballage Individually Wrapped
Sterilitet Steril
RNase fri Yes
Dnase Free Yes
Autoklaverbar Autoclavable
Form Cylindrical
Længde (metrisk) 94.0 mm
Rene krav ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
Bund form Conical (Self-standing)
Lukketype Screw Cap
Lukning inkluderet Yes
Skriveområde Yes
Uddannet Yes
Gradueringer 1 mL
Produkttype Transport Tube
Maks. RCF 12,500 x g