Få mere at vide
Troponin T (Cardiac Isoform) Ab-1 Mouse Monoclonal Antibody, Epredia™
Epredia™ Troponin T, Cardiac Isoform Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.
Brand: Epredia MS-295-P0
Beskrivelse
Troponin T is a thin filament regulatory protein of striated muscle that binds to troponin C and I and to tropomyosin and is required for calcium-dependent ATPase activity of myofibrillar proteins. It is important in proper muscle function and myofibril formation. In the human heart, four cTnT isoforms are expressed in a developmentally regulated manner. This expression in the human heart is affected by heart failure.Host Species: Mouse
Clone: 13-11
Isotype: IgG1
Species Reactivity: Human, Pig, Dog, Rabbit, Rat, Mouse, Guinea Pig, Chicken, Sparrow and Goldfish. Does not react with frog.
Epitope: FMPNLVPPKI (5' region of the cardiac gene exon 10)
Immunogen: Purified rabbit cardiac troponin T isoform (TnT4R)
Positive Control: Cardiac muscle
Cellular Localization: Mostly over the I bands in myofibrils Ab-1 specifically recognizes the cardiac isoform of Troponin T (cTnT).
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Immunofluorescence
- Electron Microscopy
Tekniske data
| Troponin T, Cardiac Isoform Ab-1 | |
| Monoclonal | |
| 200μg/mL | |
| Mouse | |
| 100 μL | |
| Muscle Biology | |
| Canine, Chicken, Goldfish, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sparrow | |
| IgG1 |
| Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 13-11 | |
| Unconjugated | |
| Purified rabbit cardiac troponin T isoform (TnT4R) | |
| IVD | |
| Primary | |
| Concentrated |
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.