missing translation for 'onlineSavingsMsg'
Få mere at vide

Epredia™ Lab Vision™ MMP-13 (Collagenase-3) Ab-1 Mouse Monoclonal Antibody with BSA and azide

Ensure accurate, reproducible results in immunohistochemistry experiments with the Epredia™ MMP-13 (Collagenase-3) Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-825-PABX

 Se flere versioner af dette produkt

Produktkode 12692407

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

Human collagenase-3 (MMP13) is a recently identified member of the matrix metalloproteinase (MMP) family that is expressed in breast carcinomas and in articular cartilage from arthritic patients. The MMP13 gene has been isolated and characterized. This gene is composed of 10 exons and 9 introns and spans over 12.5 kb. The overall organization of the collagenase-3 gene is similar to that of other MMP genes clustered at chromosome 11q22.

Host Species: Mouse

Clone: VIIIA2

Isotype: IgG1/κ

Species Reactivity: Human, Mouse and Rabbit. Others not known.

Epitope: Not determined

Immunogen: Human recombinant collagenase-3 protein

Molecular Weight: ∽60kDa (proform) and ∽48kDa (active form)

Positive Control: Conditioned, serum-free medium from (TPA-treated) human chondrosarcoma SW1353 cells; Placenta; Bladder, breast and ovarian CAs

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunoprecipitation (Native verified)
  • Affinity Purification (Not verified)

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

MMP-13 (Collagenase-3) Ab-1
Monoclonal
1mg/mL
Mouse
200 μL
Proteases And Inhibitors
Human, Mouse, Rabbit
IgG2a κ
Affinity Purification, Immunohistochemistry (Paraffin), Immunoprecipitation
VIIIA2
Unconjugated
Human recombinant collagenase-3 protein
RUO
Primary
Concentrated
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.