Få mere at vide
Epredia™ Lab Vision™ Mayer's Hematoxylin
Artikelnummer. 12603957 Shop alle Epredia produkter
Mængde:
60 ml
125 ml
Pakningsstørrelse:
125ml
60ml
Artikelnummer. 12603957
Brand: Epredia™ TA060MH
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Designed for use in histological applications, Epredia™ Lab Vision™ Mayer's Hematoxylin provides superior quality.
  • Requires slightly longer application periods
  • Based on the original formulation developed by Mayer
TRUSTED_SUSTAINABILITY
Tekniske data
Indhold og opbevaring Room temperature
Produkttype Hematoxylin
Mængde 60 ml
Til brug med (applikation) Healthcare
For in vitro diagnostic use