missing translation for 'onlineSavingsMsg'
Få mere at vide

Laminin Receptor Ab-1 Mouse Monoclonal Antibody, Epredia™

Recommended for Immunohistochemistry (Formalin/paraffin), Immunofluorescence and Flow Cytometry, Epredia™ Laminin Receptor Ab-1, Mouse Monoclonal Antibody provides accurate, reproducible results.

Brand:  Epredia MS-259-P1

 Se flere versioner af dette produkt

Produktkode 12670717

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

Laminin, one of the major glycoproteins of basement membrane, displays multiple biological activities which are mediated through its interaction with specific cell membrane receptors. Overexpression of laminin receptor is reportedly an independent prognostic factor in breast cancer and correlates with the dissemination of the tumor cells in bone marrow. Laminin receptor appears to play an important role in tumor invasion and metastasis.

Host Species: Mouse

Clone: MLuC5

Isotype: IgM

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Small cell lung carcinoma N592 live cells

Molecular Weight: 67kDa

Positive Control: Breast CA

Cellular Localization: Membrane with some cytoplasmic Ab-1 partially inhibits the binding of soluble laminin.

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Immunofluorescence
  • Flow Cytometry
  • Inhibits (partially) the binding of soluble laminin

Note:

USA: RUO; Int'l: RUO

TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

Laminin Receptor Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Extracellular Matrix
Human
IgM
Immunofluorescence, Immunohistochemistry (Paraffin)
MLuC5
Unconjugated
Small cell lung carcinoma N592 live cells
RUO
Primary
Concentrated
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.