missing translation for 'onlineSavingsMsg'
Få mere at vide

Epredia™ Lab Vision™ Factor XIII A Ab-1 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide

Ensure accurate, reproducible results in immunohistochemistry and western blotting experiments with Epredia™ Factor XIII A Ab-1, Mouse Monoclonal Antibody.

Brand:  Epredia™ MS-1237-P1

 Se flere versioner af dette produkt

Produktkode 12669346

  • ONLINE EXCLUSIVE PRICE
    3495.00 DKK
    2970.19 DKK / 500µL
    Save 524.81 DKK
    15% Slukket

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

Denne vare kan ikke returneres. Se returpolitik

Beskrivelse

Beskrivelse

Detect factor XIII in both reduced and non-reduced forms. This abtibody does not react with human Factor XIII B-chain or human Factor XII. Factor XIII is a Β-globulin found in plasma and is composed of two subunits. Factor XIII-A is the catalytic subunit and is a dimer of M.W. 160 kDa. Factor XIII is present in plasma as an alpha2Β2 heterodimer (M.W. 320 kDa); whereas in platelets, only the α2 unit exists. Factor XIIIa is a dermal dendrocyte marker and shows variable reaction with these types of tumors. It can be used for histiocytic phenotyping and has been reported to mark capillary hemangiomas and tumors of the central nervous system. Factor XIII has also been used with CD34 to differentiate between dermatofibroma and dermatofibrosarcoma protuberans.

Host Species: Mouse

Clone: AC-1A1

Isotype: IgG1/κ

Species Reactivity: Human. Others not known.

Epitope: Not determined

Immunogen: Recombinant protein corresponding to A-subunit of coagulation Factor XIII

Molecular Weight: 160 kDa (unreduced dimmer); 80 kDa (reduced monomer)

Positive Control: Recombinant human Factor XIII A-subunit; Capillary hemangioma, dermatofibroma and placenta

Cellular Localization: Cytoplasmic and nuclear

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
TRUSTED_SUSTAINABILITY
Tekniske data

Tekniske data

Factor XIII A Ab-1
Monoclonal
200μg/mL
Mouse
500 μL
Hematopoietic
Human
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot
AC-1A1
Unconjugated
Recombinant protein corresponding to A-subunit of coagulation Factor XIII
IVD
Primary
Concentrated
Produktforslag

Produktforslag

Videoer
SDS
Dokumenter

Dokumenter

Certifikater
Kampagner

Kampagner

Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.